Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647446.1 | XP_007645636.1 | 2 - 3 | 189 | 79 | |
GenBank mRNA | JI873657.1 | - | 3 - 4 | 242 | 79 | |
JP052240.1 | - | 3 - 4 | 242 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |