Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507442.2 | XP_003507490.1 | 1 - 2 | 180 | 90 | |
GenBank mRNA | JI888833.1 | - | 1 - 2 | 181 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |