Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007615073.1 | XP_007613263.1 | 5 - 6 | 682 | 65 | |
GenBank mRNA | JI887687.1 | - | 5 - 6 | 548 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |