Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497170.2 | XP_003497218.2 | 1 - 2 | 204 | 57 | |
GenBank mRNA | JI862479.1 | - | 1 - 2 | 74 | 57 | |
JP050317.1 | - | 1 - 2 | 74 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |