Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npvf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498561.2 | XP_003498609.2 | 2 - 3 | 513 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |