Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510281.2 | XP_003510329.1 | 2 - 3 | 558 | 94 | |
XM_007650918.1 | XP_007649108.1 | 2 - 3 | 548 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis 3' Most Detect Genomic DNA Probe spans exons |