Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496883.2 | XP_003496931.1 | 1 - 2 | 826 | 65 | |
XM_003496884.2 | XP_003496932.1 | 1 - 2 | 826 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |