Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514252.1 | XP_003514300.1 | 6 - 7 | 789 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |