Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siva1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498740.2 | XP_003498788.1 | 1 - 2 | 170 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Protease Probe spans exons |