Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500542.1 | XP_003500590.1 | 3 - 4 | 1417 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Detect Genomic DNA Probe spans exons |