Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496102.2 | XP_003496150.1 | 1 - 2 | 1396 | 113 | |
GenBank mRNA | JI883544.1 | - | 1 - 2 | 439 | 113 | |
JP063586.1 | - | 1 - 2 | 438 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |