Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685024.3 | XP_005685081.1 | 3 - 4 | 317 | 85 | |
GenBank mRNA | JO580841.1 | - | 2 - 3 | 668 | 85 | |
JO632256.2 | - | 2 - 3 | 449 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |