Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693300.3 | XP_005693357.1 | 2 - 3 | 670 | 78 | |
GenBank mRNA | KA319093.1 | - | 2 - 3 | 1740 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |