Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679525.3 | XP_005679582.2 | 3 - 4 | 571 | 81 | |
XM_018046866.1 | XP_017902355.1 | 3 - 4 | 483 | 81 | ||
XM_018046867.1 | XP_017902356.1 | 3 - 4 | 583 | 81 | ||
GenBank mRNA | JO515706.2 | - | 3 - 4 | 537 | 81 | |
JT688525.1 | - | 3 - 4 | 469 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |