Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684015.3 | XP_005684072.1 | 2 - 3 | 654 | 70 | |
GenBank mRNA | JT631086.1 | - | 6 - 7 | 3001 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |