Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285754.1 | NP_001272683.1 | 3 - 4 | 200 | 63 | |
GenBank mRNA | JN107562.1 | - | 3 - 4 | 200 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |