Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IYD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684915.3 | XP_005684972.2 | 1 - 2 | 245 | 69 | |
GenBank mRNA | JO438690.2 | - | 4 - 5 | 955 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |