Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041685.1 | XP_017897174.1 | 3 - 4 | 500 | 87 | |
XM_018041686.1 | XP_017897175.1 | 3 - 4 | 504 | 87 | ||
XM_018041687.1 | XP_017897176.1 | 3 - 4 | 504 | 87 | ||
GenBank mRNA | JT752954.1 | - | 3 - 4 | 379 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |