Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056039.1 | XP_017911528.1 | 4 - 5 | 2442 | 56 | |
GenBank mRNA | JT680886.1 | - | 5 - 6 | 2376 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |