Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291820.1 | NP_001278749.1 | 2 - 3 | 402 | 86 | |
GenBank mRNA | KJ136646.1 | - | 2 - 3 | 402 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |