Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287042.1 | NP_001273971.1 | 1 - 2 | 164 | 73 | |
GenBank mRNA | JO421021.2 | - | 3 - 4 | 508 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |