Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692088.3 | XP_005692145.2 | 2 - 3 | 1167 | 81 | |
XM_018062039.1 | XP_017917528.1 | 2 - 3 | 451 | 81 | ||
GenBank mRNA | JT723597.1 | - | 7 - 8 | 1613 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |