Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057426.1 | XP_017912915.1 | 3 - 4 | 702 | 68 | |
GenBank mRNA | JO579469.1 | - | 1 - 2 | 866 | 68 | |
KA341313.1 | - | 3 - 4 | 499 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |