Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685411.3 | XP_005685468.2 | 3 - 4 | 1803 | 63 | |
XM_018054513.1 | XP_017910002.1 | 3 - 4 | 1755 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |