Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044301.1 | XP_017899790.1 | 3 | 1251 | 77 | |
XM_018044302.1 | XP_017899791.1 | 3 | 537 | 77 | ||
GenBank mRNA | JO458964.2 | - | 1 | 242 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |