Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973931.2 | XP_013829385.1 | 4 - 5 | 1229 | 99 | |
XM_018039266.1 | XP_017894755.1 | 4 - 5 | 1226 | 99 | ||
XM_018039267.1 | XP_017894756.1 | 4 - 5 | 1230 | 99 | ||
GenBank mRNA | KA327333.1 | - | 4 - 5 | 1267 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |