Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013968196.2 | XP_013823650.2 | 2 - 3 | 723 | 100 | |
XM_018056541.1 | XP_017912030.1 | 2 - 3 | 723 | 100 | ||
XM_018056542.1 | XP_017912031.1 | 3 - 4 | 451 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |