Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678910.3 | XP_005678967.2 | 2 - 3 | 2219 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |