Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677743.3 | XP_005677800.2 | 13 - 14 | 3150 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |