Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038742.1 | XP_017894231.1 | 8 - 9 | 3919 | 93 | |
XM_018038743.1 | XP_017894232.1 | 8 - 9 | 2954 | 93 | ||
XM_018038744.1 | XP_017894233.1 | 8 - 9 | 2969 | 93 | ||
XM_018038745.1 | XP_017894234.1 | 7 - 8 | 2889 | 93 | ||
XM_018038746.1 | XP_017894235.1 | 7 - 8 | 3677 | 93 | ||
XM_018038747.1 | XP_017894236.1 | 7 - 8 | 3419 | 93 | ||
XM_018038748.1 | XP_017894237.1 | 7 - 8 | 3113 | 93 | ||
XM_018038749.1 | XP_017894238.1 | 6 - 7 | 2970 | 93 | ||
XM_018038750.1 | XP_017894239.1 | 7 - 8 | 2826 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |