Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674692.3 | XP_005674749.2 | 3 - 4 | 1129 | 140 | |
GenBank mRNA | JO353076.2 | - | 4 - 5 | 430 | 140 | |
JO375642.2 | - | 1 - 2 | 55 | 140 | ||
JO415835.2 | - | 4 - 5 | 433 | 140 | ||
JO437008.2 | - | 3 - 4 | 625 | 140 | ||
JO504230.2 | - | 3 - 4 | 537 | 140 | ||
JO551203.2 | - | 4 - 5 | 416 | 140 | ||
JO594966.2 | - | 3 - 4 | 617 | 140 | ||
JO631207.2 | - | 3 - 4 | 579 | 140 | ||
JT659292.1 | - | 3 - 4 | 132 | |||
JT717661.1 | - | 3 - 4 | 423 | 140 | ||
KA321950.1 | - | 3 - 4 | 712 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |