Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO437008.2 | - | 4 - 5 | 773 | 96 | |
JO504230.2 | - | 4 - 5 | 685 | 96 | ||
JO551203.2 | - | 3 - 4 | 268 | 96 | ||
JO594966.2 | - | 4 - 5 | 765 | 96 | ||
JO631207.2 | - | 4 - 5 | 727 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |