Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038287.1 | XP_017893776.1 | 3 - 4 | 526 | 125 | |
GenBank mRNA | JO440171.2 | - | 3 - 4 | 492 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |