Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684903.3 | XP_005684960.1 | 5 - 6 | 588 | 68 | |
XM_005684904.3 | XP_005684961.1 | 3 - 4 | 391 | 68 | ||
GenBank mRNA | JT704540.1 | - | 6 - 7 | 1326 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |