Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690983.3 | XP_005691040.1 | 10 - 11 | 1104 | 130 | |
XM_005690986.2 | XP_005691043.1 | 9 - 10 | 1008 | 130 | ||
XM_005690987.2 | XP_005691044.1 | 9 - 10 | 982 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |