Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052670.1 | XP_017908159.1 | 4 - 5 | 494 | 74 | |
XM_018052671.1 | XP_017908160.1 | 4 - 5 | 494 | 74 | ||
XM_018052672.1 | XP_017908161.1 | 3 - 4 | 407 | 74 | ||
XM_018052673.1 | XP_017908162.1 | 4 - 5 | 494 | 74 | ||
XM_018052674.1 | XP_017908163.1 | 4 - 5 | 494 | 74 | ||
GenBank mRNA | KA314190.1 | - | 4 - 5 | 459 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |