Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039254.1 | XP_017894743.1 | 2 - 3 | 619 | 108 | |
GenBank mRNA | KA328079.1 | - | 3 - 4 | 2623 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |