Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPEPLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679103.3 | XP_005679160.1 | 2 - 3 | 478 | 87 | |
XM_013963249.2 | XP_013818703.2 | 2 - 3 | 944 | 87 | ||
GenBank mRNA | JO499546.2 | - | 2 - 3 | 352 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |