Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051309.1 | XP_017906798.1 | 9 - 10 | 4593 | 80 | |
XM_018051310.1 | XP_017906799.1 | 8 - 9 | 4422 | 80 | ||
XM_018051311.1 | XP_017906800.1 | 8 - 9 | 4194 | 80 | ||
XM_018051312.1 | XP_017906801.1 | 7 - 8 | 4023 | 80 | ||
XM_018051313.1 | XP_017906802.1 | 7 - 8 | 2352 | 80 | ||
XM_018051314.1 | XP_017906803.1 | 5 - 6 | 1782 | 80 | ||
XM_018051315.1 | XP_017906804.1 | 4 - 5 | 1314 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |