Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013969300.2 | XP_013824754.2 | 3 - 4 | 2571 | 109 | |
GenBank mRNA | JT683129.1 | - | 3 - 4 | 1482 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |