Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061864.1 | XP_017917353.1 | 2 - 3 | 279 | 75 | |
XM_018061865.1 | XP_017917354.1 | 2 - 3 | 220 | 75 | ||
GenBank mRNA | JO578964.1 | - | 2 - 3 | 324 | 75 | |
JO643282.2 | - | 2 - 3 | 423 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |