Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF38B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962575.2 | XP_013818029.1 | 1 - 2 | 558 | 62 | |
GenBank mRNA | JO368008.2 | - | 5 - 6 | 1642 | 62 | |
JO579267.1 | - | 5 - 6 | 2149 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |