Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057935.1 | XP_017913424.1 | 1 - 2 | 478 | 74 | |
GenBank mRNA | JO590180.2 | - | 1 - 2 | 311 | 74 | |
KA325673.1 | - | 1 - 2 | 349 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |