Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678295.3 | XP_005678352.2 | 1 - 2 | 196 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |