Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042095.1 | XP_017897584.1 | 6 - 7 | 1551 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |