Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173061.1 | NP_001166532.1 | 16 - 17 | 2497 | 83 | |
GenBank mRNA | D84339.1 | - | 16 - 17 | 2497 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Cavia porcellus Probe spans exons |