Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172970.1 | NP_001166441.1 | 3 - 4 | 340 | 108 | |
GenBank mRNA | U34588.1 | - | 3 - 4 | 340 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |