Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: loqs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298965.1 | NP_001285894.1 | 3 - 4 | 1187 | 96 | |
NM_135802.4 | NP_609646.1 | 3 - 4 | 1187 | 96 | ||
GenBank mRNA | AY071070.1 | - | 3 - 4 | 1184 | 96 | |
DQ118034.1 | - | 3 - 4 | 1191 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |