Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: siz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043153.2 | NP_001036618.1 | 3 - 4 | 1285 | 106 | |
NM_001170016.1 | NP_001163487.1 | 3 - 4 | 1562 | 106 | ||
NM_168881.3 | NP_730594.1 | 3 - 4 | 1761 | 106 | ||
NM_206407.2 | NP_996129.1 | 4 - 5 | 1945 | 106 | ||
GenBank mRNA | AY375487.1 | - | 3 - 4 | 1757 | 106 | |
AY375488.1 | - | 3 - 4 | 1705 | 106 | ||
AY375489.1 | - | 3 - 4 | 1285 | 106 | ||
BT150471.1 | - | 3 - 4 | 1757 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |