Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079213.4 | NP_523937.2 | 6 - 7 | 4258 | 67 | |
NM_168129.4 | NP_729087.1 | 6 - 7 | 4431 | 67 | ||
NM_206279.4 | NP_996001.2 | 8 - 9 | 4449 | 67 | ||
GenBank mRNA | BT003784.1 | - | 6 - 7 | 4411 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |